Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
BAM7 ≥98%

BAM7 ≥98%

Bottle
Katalog # APOSBUP10591-100MG
New Product
Leverandør:   Apollo Scientific
BAM7 ≥98%
Katalog # APOSBUP10591-100MG
Leverandør:  Apollo Scientific
New Product
CAS nummer:  331244-89-4
BAM7 ≥98%

Bottle

CAS nummer:  331244-89-4
Price
3.440,00 kr
Log ind for tilgængelighed
1 * 100 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonymer:  3-Methyl-1-(4-phenyl-2-thiazolyl)-1H-pyrazole-4,5-dione 4-[2-(2-ethoxyphenyl)hydrazone], 4-(2-(2-Ethoxyphenyl)hydrazono)-5-methyl-2-(4-phenylthiazol-2-yl)-2,4-dihydro-3H-pyrazol-3-one, Bax Activator Molecule 7
  • CAS nummer:331244-89-4
  • Formel:C₂₁H₁₉N₅O₂S
  • Molecular Weight:405,47 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.