AC-55541 ≥98%
Bottle
Katalog # APOSBUP09838-25MG
New Product
Leverandør: Apollo Scientific
AC-55541 ≥98%
Katalog # APOSBUP09838-25MG
Leverandør: Apollo Scientific
New Product
CAS nummer: 916170-19-9
Gå til:
Product Details & Documents
Usage: Biochemical/life science applications.
Synonymer: (E)-N-(2-(2-(1-(3-Bromophenyl)ethylidene)hydrazineyl)-2-oxo-1-(4-oxo-3,4-dihydrophthalazin-1-yl)ethyl)benzamide
- CAS nummer:916170-19-9
- Formel:C₂₅H₂₀BrN₅O₃
- Molecular Weight:518,36 g/mol
Specifikationer
Specifikationer
- Pakning:Bottle



