Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Dibutyl phthalate ≥98%

Dibutyl phthalate ≥98%

Bottle
Katalog # APOSBUP17288-5G
New Product
Leverandør:   Apollo Scientific
Dibutyl phthalate ≥98%
Katalog # APOSBUP17288-5G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  84-74-2
Dibutyl phthalate ≥98%

Bottle

CAS nummer:  84-74-2
Price
4.270,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonymer:  DBP
  • CAS nummer:84-74-2
  • Formel:C₁₆H₂₂O₄
  • Molecular Weight:278,34 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.