Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
DEPC ≥98%

DEPC ≥98%

Bottle
Katalog # APOSBUP17265-5G
New Product
Leverandør:   Apollo Scientific
DEPC ≥98%
Katalog # APOSBUP17265-5G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  1609-47-8
DEPC ≥98%

Bottle

CAS nummer:  1609-47-8
Price
2.070,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents

Usage: Biochemical/life science applications

Synonymer:  DEPC (Diethylpyrocarbonate), Diethyl dicarbonate, DEPC
  • CAS nummer:1609-47-8
  • Formel:C₆H₁₀O₅
  • Molecular Weight:162,14 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.