DEPC ≥98%
Bottle
Katalog # APOSBUP17265-500MG
New Product
Leverandør: Apollo Scientific
DEPC ≥98%
Katalog # APOSBUP17265-500MG
Leverandør: Apollo Scientific
New Product
CAS nummer: 1609-47-8
Gå til:
Product Details & Documents
Usage: Biochemical/life science applications
Synonymer: DEPC (Diethylpyrocarbonate), Diethyl dicarbonate, DEPC
- CAS nummer:1609-47-8
- Formel:C₆H₁₀O₅
- Molecular Weight:162,14 g/mol
Specifikationer
Specifikationer
- Pakning:Bottle