Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Oleic acid ≥95%

Bottle
Catalog # APOSBUP18491-5G
New Product
Supplier:   Apollo Scientific
Oleic acid ≥95%
Catalog # APOSBUP18491-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  112-80-1
Oleic acid ≥95%

Bottle

CAS Number:  112-80-1
Price
845,00 kr
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  (9Z)-9-Octadecenoic acid
  • CAS Number:112-80-1
  • Formula:C₁₈H₃₄O₂
  • Molecular Weight:282,46 g/mol
Specifications

Specifications

  • Pakning:
    Bottle

Specifications

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.