Oleic acid ≥95%
Bottle
Katalog # APOSBUP18491-1G
New Product
Leverandør: Apollo Scientific
Oleic acid ≥95%
Katalog # APOSBUP18491-1G
Leverandør: Apollo Scientific
New Product
CAS nummer: 112-80-1
Gå til:
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonymer: (9Z)-9-Octadecenoic acid
- CAS nummer:112-80-1
- Formel:C₁₈H₃₄O₂
- Molecular Weight:282,46 g/mol
Specifikationer
Specifikationer
- Pakning:Bottle
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



