Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Colchicine ≥98%

Colchicine ≥98%

Bottle
Katalog # APOSBUP16899-500MG
New Product
Leverandør:   Apollo Scientific
Colchicine ≥98%
Katalog # APOSBUP16899-500MG
Leverandør:  Apollo Scientific
New Product
CAS nummer:  64-86-8
Colchicine ≥98%

Bottle

CAS nummer:  64-86-8
Price
626,00 kr
Log ind for tilgængelighed
1 * 500 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonymer:  7α-H-Colchicine, (S)-(-)-Colchicine
  • CAS nummer:64-86-8
  • Formel:C₂₂H₂₅NO₆
  • Molecular Weight:399,44 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.