Crystal Violet ≥90%
Bottle
Katalog # APOSBUP18043-10G
New Product
Leverandør: Apollo Scientific
Crystal Violet ≥90%
Katalog # APOSBUP18043-10G
Leverandør: Apollo Scientific
New Product
CAS nummer: 548-62-9
Gå til:
Product Details & Documents
Usage: Biochemical/life science applications
Synonymer: 4-[4,4'-Bis(dimethylamino)benzhydryliden]cyclohexa-2,5-dien-1-yliden]dimethylammoniumchlorid, CI 42555
- CAS nummer:548-62-9
- Formel:C₂₅H₃₀ClN₃
- Molecular Weight:407,98 g/mol
- EINECS:208-953-6
Specifikationer
Specifikationer
- Pakning:Bottle



