Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Crystal Violet ≥90%

Crystal Violet ≥90%

Bottle
Katalog # APOSBUP18043-5G
Irriterende
Sundhed
New Product
Leverandør:   Apollo Scientific
Crystal Violet ≥90%
Katalog # APOSBUP18043-5G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  548-62-9
Crystal Violet ≥90%

Bottle

CAS nummer:  548-62-9
Price
593,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents

Usage: Biochemical/life science applications

Synonymer:  4-[4,4'-Bis(dimethylamino)benzhydryliden]cyclohexa-2,5-dien-1-yliden]dimethylammoniumchlorid, CI 42555
  • CAS nummer:548-62-9
  • Formel:C₂₅H₃₀ClN₃
  • Molecular Weight:407,98 g/mol
  • EINECS:208-953-6
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.