Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Cinnamaldehyde ≥80%

Cinnamaldehyde ≥80%

Bottle
Katalog # APOSBUP17096-100MG
Irriterende
New Product
Leverandør:   Apollo Scientific
Cinnamaldehyde ≥80%
Katalog # APOSBUP17096-100MG
Leverandør:  Apollo Scientific
New Product
CAS nummer:  104-55-2
Cinnamaldehyde ≥80%

Bottle

CAS nummer:  104-55-2
Price
297,00 kr
Log ind for tilgængelighed
1 * 100 mg
Product Details & Documents

Usage: Biochemical/life science applications.

Synonymer:  trans-Cinnamaldehyde, 3-Phenylacrylaldehyde
  • CAS nummer:104-55-2
  • Formel:C₉H₈O
  • MDL nummer:MFCD00007000
  • Molecular Weight:132,16 g/mol
  • EINECS:203-213-9
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer