Order Entry
Denmark
Orders LinkContactUsLinkComponent
Docusate sodium ≥98%

Docusate sodium ≥98%

Bottle
Katalog # APOSBUP17054-100MG
New Product
Leverandør:   Apollo Scientific
Docusate sodium ≥98%
Katalog # APOSBUP17054-100MG
Leverandør:  Apollo Scientific
New Product
CAS nummer:  577-11-7
Docusate sodium ≥98%

Bottle

CAS nummer:  577-11-7
Price
483,00 kr
Log ind for tilgængelighed
1 * 100 mg
Product Details & Documents

Usage: Biochemical/life science applications.

Synonymer:  Dioctyl sulphosuccinate sodium salt, Dioctyl sulphosuccinate, sodium salt, Sodium dioctyl sulphosuccinate, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulphonate, bis(2-Ethylhexyl) sodium sulphosuccinate
  • CAS nummer:577-11-7
  • Formel:C₂₀H₃₇NaO₇S
  • Molecular Weight:444,56 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer