Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Choline chloride ≥95%

Bottle
  APOSBUP17045-10G
New Product
Choline chloride ≥95%
  APOSBUP17045-10G
New Product
 :  67-48-1
Choline chloride ≥95%

Bottle

 :  67-48-1
 
4.340,00 kr
   
1 * 10 g
 

Usage: Biochemical/life science applications

 :  2-hydroxy-N,N,N-trimethylethanaminium chloride
  •  :67-48-1
  •  :C₅H₁₄ClNO
  •  :139,63 g/mol
 

 

  • Pakning:
    Bottle