Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Choline chloride ≥95%

Choline chloride ≥95%

Bottle
Catalog # APOSBUP17045-500MG
New Product
Supplier:   Apollo Scientific
Choline chloride ≥95%
Catalog # APOSBUP17045-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  67-48-1
Choline chloride ≥95%

Bottle

CAS Number:  67-48-1
Price
922,00 kr
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  2-hydroxy-N,N,N-trimethylethanaminium chloride
  • CAS Number:67-48-1
  • Formula:C₅H₁₄ClNO
  • Molecular Weight:139,63 g/mol
Specifications

Specifications

  • Pakning:
    Bottle

Specifications