Orders LinkContactUsLinkComponent
Torkinib ≥98%
Torkinib ≥98%
Bottle
  APOSBUP15517-50MG
New Product
Torkinib ≥98%
  APOSBUP15517-50MG
New Product
 :  1092351-67-1
Torkinib ≥98%
Bottle
 :  1092351-67-1
1.340,00 kr
1 * 50 mg
Usage: Biochemical/ life science applications.
 :  2-(4-Amino-1-isopropyl-1H-pyrazolo[3,4-d]pyrimidin-3-yl)-1H-indol-5-ol, Torkinib, 2-[4-Amino-1-(1-methylethyl)-1H-pyrazolo[3,4-d]pyrimidin-3-yl]-1H-indol-5-ol
  •  :1092351-67-1
  •  :C₁₆H₁₆N₆O
  •  :308,34 g/mol
  • Pakning:
    Bottle