Methoxyfenozide ≥98%
Bottle
Catalog # APOSBUP16836-5G
New Product
Supplier: Apollo Scientific
Methoxyfenozide ≥98%
Catalog # APOSBUP16836-5G
Supplier: Apollo Scientific
New Product
CAS Number: 161050-58-4
Jump to:
Product Details & Documents
Usage: Biochemical/life science applications.
Synonyms: Methoxyphenozide
- CAS Number:161050-58-4
- Formula:C₂₂H₂₈N₂O₃
- Molecular Weight:368,47 g/mol
Specifications
Specifications
- Pakning:Bottle