Orders LinkContactUsLinkComponent
Methoxyfenozide ≥98%
Methoxyfenozide ≥98%
Bottle
Katalog # APOSBUP16836-500MG
New Product
Leverandør:   Apollo Scientific
Methoxyfenozide ≥98%
Katalog # APOSBUP16836-500MG
Leverandør:  Apollo Scientific
New Product
CAS nummer:  161050-58-4
Methoxyfenozide ≥98%
Bottle
CAS nummer:  161050-58-4
Price
1.350,00 kr
Log ind for tilgængelighed
1 * 500 mg
Product Details & Documents
Usage: Biochemical/life science applications.
Synonymer:  Methoxyphenozide
  • CAS nummer:161050-58-4
  • Formel:C₂₂H₂₈N₂O₃
  • Molecular Weight:368,47 g/mol
Specifikationer
Specifikationer
  • Pakning:
    Bottle