Methoxyfenozide ≥98%
Bottle
Katalog # APOSBUP16836-100MG
New Product
Leverandør: Apollo Scientific
Methoxyfenozide ≥98%
Katalog # APOSBUP16836-100MG
Leverandør: Apollo Scientific
New Product
CAS nummer: 161050-58-4
Gå til:
Product Details & Documents
Usage: Biochemical/life science applications.
Synonymer: Methoxyphenozide
- CAS nummer:161050-58-4
- Formel:C₂₂H₂₈N₂O₃
- Molecular Weight:368,47 g/mol
Specifikationer
Specifikationer
- Pakning:Bottle