Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Hyperoside ≥98%

Bottle
  APOSBUP15509-200MG
New Product
Hyperoside ≥98%
  APOSBUP15509-200MG
New Product
 :  482-36-0
Hyperoside ≥98%

Bottle

 :  482-36-0
 
4.520,00 kr
   
1 * 200 mg
 

Usage: Biochemical/life science applications

 :  Hyperoside, 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-3-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one, Hyperin, Quercetin-3-galactoside
  •  :482-36-0
  •  :C₂₁H₂₀O₁₂
  •  :464,38 g/mol
 

 

  • Pakning:
    Bottle

 

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.