2-Chlorohexane 98%
Bottle
Katalog # APOSOR1023669-5G
New Product
Leverandør: Apollo Scientific
2-Chlorohexane 98%
Katalog # APOSOR1023669-5G
Leverandør: Apollo Scientific
New Product
CAS nummer: 638-28-8
Gå til:
Product Details & Documents
Usage: Organic building block/reagent.
Synonymer: 2-Hexyl chloride
- CAS nummer:638-28-8
- Formel:C₆H₁₃Cl
- Molecular Weight:120,62 g/mol
Specifikationer
Specifikationer
- Pakning:Bottle



