Methotrexate ≥95%
Bottle
Katalog # APOSBUP17033-100MG
New Product
Leverandør: Apollo Scientific
Methotrexate ≥95%
Katalog # APOSBUP17033-100MG
Leverandør: Apollo Scientific
New Product
CAS nummer: 59-05-2
Gå til:
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonymer: (S)-2-(4-(((2,4-Diaminopteridin-6-yl)methyl)(methyl)amino)benzamido)pentanedioic acid
- CAS nummer:59-05-2
- Formel:C₂₀H₂₂N₈O₅
- Molecular Weight:454,44 g/mol
Specifikationer
Specifikationer
- Pakning:Bottle
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



