Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Silica 99, 8%, fumed

Bottle
  APOSOR1024457-2.5K
 
New Product
Silica 99, 8%, fumed
  APOSOR1024457-2.5K
New Product
 :  112945-52-5
Silica 99, 8%, fumed

Bottle

 :  112945-52-5
 
1.420,00 kr
   
1 * 2,5 kg
 

Hydrophobic type R974, Specific surface area 170 ±20 m²/g; Particle size: 12 nm.

  • Usage: Organic building block/reagent
  •  :112945-52-5
  •  :SiO₂
  •  :60,08 g/mol
 

 

  • Pakning:
    Bottle

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.