Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Potassium Polysulphide 42%

Potassium Polysulphide 42%

Bottle
Katalog # APOSOR1028673-5G
New Product
Leverandør:   Apollo Scientific
Potassium Polysulphide 42%
Katalog # APOSOR1028673-5G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  37199-66-9
Potassium Polysulphide 42%

Bottle

CAS nummer:  37199-66-9
Price
165,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents

Usage: Organic building block/reagent

Synonymer:  Potassium polysulphide, Liver of sulphur, Sulphurated potash
  • CAS nummer:37199-66-9
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.