Hyperoside 98%
Bottle
Katalog # APOSOR1023063-100M
New Product
Leverandør: Apollo Scientific
Hyperoside 98%
Katalog # APOSOR1023063-100M
Leverandør: Apollo Scientific
New Product
CAS nummer: 482-36-0
Gå til:
Product Details & Documents
Usage: Organic building block/reagent.
Synonymer: Hyperoside, 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-3-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one, Hyperin, Quercetin-3-galactoside
- CAS nummer:482-36-0
- Formel:C₂₁H₂₀O₁₂
- Molecular Weight:464,38 g/mol
Specifikationer
Specifikationer
- Pakning:Bottle



