Sulfanilamid
Bottle
Katalog # APOSBIMB1014-25G
New Product
Leverandør: Apollo Scientific
Gå til:
Product Details & Documents
Synonymer: 4-Aminobenzenesulphonamide
- CAS nummer:63-74-1
- Formel:H₂NC₆H₄SO₂NH₂
- MDL nummer:MFCD00007939
- Molecular Weight:172.21 g/mol
- Opbevaringstemperatur:Stue temperatur
Specifikationer
Specifikationer
- Pakning:Bottle
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



