Natriumpyruvat
Bottle
Katalog # APOSBIB6270-100G
New Product
Leverandør: Apollo Scientific
Natriumpyruvat
Katalog # APOSBIB6270-100G
Leverandør: Apollo Scientific
New Product
CAS nummer: 113-24-6
Gå til:
Product Details & Documents
Medium of animal cell culture; Material for pharmaceuticals, agrochemicals and diagnostic reagent; Intermediate for flavors and fragrances.
Synonymer: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
- CAS nummer:113-24-6
- Formel:CH₃COCOONa
- MDL nummer:MFCD00002586
- Molecular Weight:110.04 g/mol
- Smeltende Pt:>300 °C
- Smeltende Pt:>300 °C
- Opbevaringstemperatur:Køleskab
Specifikationer
Specifikationer
- Pakning:Bottle
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



