Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Cyclobutanol

Cyclobutanol

Bottle
Katalog # APOSOR965150-1G
New Product
Leverandør:   Apollo Scientific
Cyclobutanol
Katalog # APOSOR965150-1G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  2919-23-5
Cyclobutanol

Bottle

CAS nummer:  2919-23-5
Price
165,00 kr
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents
  • CAS nummer:2919-23-5
  • Formel:C₄H₈O
  • MDL nummer:MFCD00001318
  • Molecular Weight:72.11 g/mol
  • Flash Pt:21 °C
  • Opbevaringstemperatur:Stue temperatur
  • FN:1987
  • ADR:3,II
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.