Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Imidazol 99+%

Imidazol 99+%

Bottle
Catalog # APOSBI7565-2.5KG
Corrosive
Irritant
Health
New Product
Supplier:   Apollo Scientific
Imidazol 99+%
Catalog # APOSBI7565-2.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  288-32-4
Imidazol 99+%

Bottle

CAS Number:  288-32-4
Price
1.930,00 kr
Log in for availability
1 * 2,5 kg
Product Details & Documents

Biological buffer.

  • Assay (by titration) 99+%
  • Water (by Karl Fischer) <1.0%
  • White crystals
Synonyms:  Glyoxaline, 1H-Imidazole, Glyoxalamine
  • CAS Number:288-32-4
  • Formula:C₃H₄N₂
  • MDL Number:MFCD00005183
  • Molecular Weight:68.08 g/mol
  • Boiling Pt:235 °C
  • Melting Pt:85…91 °C
  • Flash Pt:145 °C
  • Storage Temperature:Stue temperatur
  • EINECS:206-019-2
  • UN:3263
  • ADR:8,II
Specifications

Specifications

  • Pakning:
    Bottle

Specifications