Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Imidazol 99+%

Imidazol 99+%

Bottle
Katalog # APOSBI7565-100G
Ætsende
Irriterende
Sundhed
New Product
Leverandør:   Apollo Scientific
Imidazol 99+%
Katalog # APOSBI7565-100G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  288-32-4
Imidazol 99+%

Bottle

CAS nummer:  288-32-4
Price
187,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Biological buffer.

  • Assay (by titration) 99+%
  • Water (by Karl Fischer) <1.0%
  • White crystals
Synonymer:  Glyoxaline, 1H-Imidazole, Glyoxalamine
  • CAS nummer:288-32-4
  • Formel:C₃H₄N₂
  • MDL nummer:MFCD00005183
  • Molecular Weight:68.08 g/mol
  • Kogende Pt:235 °C
  • Smeltende Pt:85…91 °C
  • Flash Pt:145 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:206-019-2
  • FN:3263
  • ADR:8,II
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.