Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
(S)-(-)-2-Acetoxypropionic acid

(S)-(-)-2-Acetoxypropionic acid

Bottle
Katalog # APOSOR322114-100G
Irriterende
New Product
Leverandør:   Apollo Scientific
(S)-(-)-2-Acetoxypropionic acid
Katalog # APOSOR322114-100G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  6034-46-4
(S)-(-)-2-Acetoxypropionic acid

Bottle

CAS nummer:  6034-46-4
Price
6.810,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
Synonymer:  (-)-O-Acetyl-L-lactic Acid, (S)-2-Acetoxypropanoic acid
  • CAS nummer:6034-46-4
  • Formel:C₅H₈O₄
  • MDL nummer:MFCD00191370
  • Molecular Weight:132.12 g/mol
  • Kogende Pt:142 °C/20mmHg
  • Tæthed:1,18 g/cm³ (25 °C)
  • Opbevaringstemperatur:Stue temperatur
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.