Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

3,5-Dihydroxybenzonitril 98%

Bottle
Katalog # APOSOR5346-250MG
New Product
Leverandør:   Apollo Scientific
3,5-Dihydroxybenzonitril 98%
Katalog # APOSOR5346-250MG
Leverandør:  Apollo Scientific
New Product
CAS nummer:  19179-36-3
3,5-Dihydroxybenzonitril 98%

Bottle

CAS nummer:  19179-36-3
Price
165,00 kr
Log ind for tilgængelighed
1 * 250 mg
Product Details & Documents
  • CAS nummer:19179-36-3
  • Formel:C₇H₅NO₂
  • MDL nummer:MFCD00016453
  • Molecular Weight:135.12 g/mol
  • Smeltende Pt:185…190 °C
  • Opbevaringstemperatur:Stue temperatur
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.