Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
3,5-Dihydroxybenzonitril 98%

3,5-Dihydroxybenzonitril 98%

Bottle
Catalog # APOSOR5346-100G
New Product
Supplier:   Apollo Scientific
3,5-Dihydroxybenzonitril 98%
Catalog # APOSOR5346-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  19179-36-3
3,5-Dihydroxybenzonitril 98%

Bottle

CAS Number:  19179-36-3
Price
9.750,00 kr
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:19179-36-3
  • Formula:C₇H₅NO₂
  • MDL Number:MFCD00016453
  • Molecular Weight:135.12 g/mol
  • Melting Pt:185…190 °C
  • Storage Temperature:Stue temperatur
Specifications

Specifications

  • Pakning:
    Bottle

Specifications

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.