Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
beta-Alanine 99%

beta-Alanine 99%

Bottle
Katalog # APOSOR5225-100G
New Product
Leverandør:   Apollo Scientific
beta-Alanine 99%
Katalog # APOSOR5225-100G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  107-95-9
beta-Alanine 99%

Bottle

CAS nummer:  107-95-9
Price
396,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Usage: Organic building block/reagent

  • Non-hazardous
Synonymer:  H-β-Ala-OH, 3-Aminopropionic acid, H-Bala-OH, 3-Aminopropanoic acid
  • CAS nummer:107-95-9
  • Formel:C₃H₇NO₂
  • MDL nummer:MFCD00008200
  • Molecular Weight:89,09318 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.