Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
N-Boc-D-prolinal

N-Boc-D-prolinal

Bottle
Katalog # APOSOR316011-500G
New Product
Leverandør:   Apollo Scientific
N-Boc-D-prolinal
Katalog # APOSOR316011-500G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  73365-02-3
N-Boc-D-prolinal

Bottle

CAS nummer:  73365-02-3
Price
18.700,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents
Synonymer:  1-Pyrrolidinecarboxylic acid, 2-formyl-, 1,1-dimethylethyl ester, (2R)-, tert-Butyl (R)-2-Formylpyrrolidine-1-carboxylate, (R)-N-BOC-Prolinal, (R)-2-Formylpyrrolidine-1-carboxylic acid tert-Butyl ester, N-Boc-D-Prolinal, N-Boc-D-prolinal, N-(tert-Butoxycarbonyl)-D-prolinal, (R)-tert-butyl 2-formylpyrrolidine-1-carboxylate
  • CAS nummer:73365-02-3
  • Formel:C₁₀H₁₇NO₃
  • MDL nummer:MFCD01321389
  • Molecular Weight:199.25 g/mol
  • Opbevaringstemperatur:Stue temperatur
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer