(3,5-Dichlorophenyl)acetic acid
Bottle
Katalog # APOSOR51794-500G
New Product
Leverandør: Apollo Scientific
(3,5-Dichlorophenyl)acetic acid
Katalog # APOSOR51794-500G
Leverandør: Apollo Scientific
New Product
CAS nummer: 51719-65-4
Gå til:
Product Details & Documents
Synonymer: 2-(3,5-Dichlorophenyl)acetic acid
- CAS nummer:51719-65-4
- Formel:C₈H₆Cl₂O₂
- MDL nummer:MFCD01764671
- Molecular Weight:205.03 g/mol
- Opbevaringstemperatur:Fryser
Specifikationer
Specifikationer
- Pakning:Bottle



