Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
(3,5-Dichlorophenyl)acetic acid

(3,5-Dichlorophenyl)acetic acid

Bottle
Katalog # APOSOR51794-100G
Irriterende
New Product
Leverandør:   Apollo Scientific
(3,5-Dichlorophenyl)acetic acid
Katalog # APOSOR51794-100G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  51719-65-4
(3,5-Dichlorophenyl)acetic acid

Bottle

CAS nummer:  51719-65-4
Price
3.850,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
Synonymer:  2-(3,5-Dichlorophenyl)acetic acid
  • CAS nummer:51719-65-4
  • Formel:C₈H₆Cl₂O₂
  • MDL nummer:MFCD01764671
  • Molecular Weight:205.03 g/mol
  • Opbevaringstemperatur:Fryser
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.