Temperature Data Loggers
AISI-316L stainless steel disk temperature recorder DLDISK3618_CL, 80 g, for accurate and precise temperature monitoring; External (Ø×H): 36×18 mm
Catalog # ESPIDLDISK3618_CL
New Product
Supplier: RAYPA
Jump to:
Product Details & Documents
AISI-316L stainless steel disk temperature recorder with connection base and software.
- User-friendly operation
- Data logger available in multiple sizes to suit different needs
- Delivers high accuracy and precision
Recommended for autoclave validation and for monitoring the internal temperature of containers.
Specifications
- Beskrivelse:AISI-316L stainless steel disk temperature recorder DLDISK3618_CL, 80 g, for accurate and precise temperature monitoring; External (Ø×H): 36×18 mm
- Seq:2