Orders LinkContactUsLinkComponent
Temperature Data Loggers
Siden oplever i øjeblikket tekniske problemer. Prøv venligst igen senere. Vi beklager ulejligheden.
Temperature Data Loggers
AISI-316L stainless steel disk temperature recorder with connection base and software BDL-DISK3618_CL, 200 g, for autoclave validation and for monitoring the internal temperature of containers, 60×95×175 mm (External)
Katalog # ESPIBDL-DISK3618_C
New Product
Leverandør:   RAYPA
Temperature Data Loggers
Katalog # ESPIBDL-DISK3618_C
Leverandør:  RAYPA
New Product
Temperature Data Loggers
Temperature Data Loggers
AISI-316L stainless steel disk temperature recorder with connection base and software BDL-DISK3618_CL, 200 g, for autoclave validation and for monitoring the internal temperature of containers, 60×95×175 mm (External)
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    AISI-316L stainless steel disk temperature recorder with connection base and software BDL-DISK3618_CL, 200 g, for autoclave validation and for monitoring the internal temperature of containers
  • Seq:
    3
  • B×D×H:
    60×95×175 mm (External)