Temperature Data Loggers
AISI-316L stainless steel disk temperature recorder with connection base and software, BDL-DISK3618_TL, 200 g, for autoclave validation and for monitoring the internal temperature of containers, 60×95×175 mm (External)
Katalog # ESPIBDL-DISK3618_T
New Product
Leverandør: RAYPA
Gå til:
Product Details & Documents
AISI-316L stainless steel disk temperature recorder with connection base and software.
- User-friendly operation
- Data logger available in multiple sizes to suit different needs
- Delivers high accuracy and precision
Recommended for autoclave validation and for monitoring the internal temperature of containers.
Specifikationer
- Beskrivelse:AISI-316L stainless steel disk temperature recorder with connection base and software, BDL-DISK3618_TL, 200 g, for autoclave validation and for monitoring the internal temperature of containers
- Seq:4
- B×D×H:60×95×175 mm (External)