VWR® Socket Caps and Colour Inserts for Screw Microtubes
Customisable screw caps, non sterile
Catalog # 525-1649
New Product
Supplier: VWR Collection
VWR® Socket Caps and Colour Inserts for Screw Microtubes
Catalog # 525-1649
Supplier: VWR Collection
New Product
Jump to:
Product Details & Documents
Customisable socket caps with O-ring, accept colourful cap inserts to organise samples or match kit branding. For use with VWR® screw cap microtubes only.
Feature ¾ turn to easily open and close tubes requiring less revolutions than competitor tubes.
Specifications
- Beskrivelse:Customisable screw caps, non sterile
- Seq:2