Orders LinkContactUsLinkComponent
VWR® Socket Caps and Colour Inserts for Screw Microtubes
VWR® Socket Caps and Colour Inserts for Screw Microtubes
Customisable screw caps, non sterile
Katalog # 525-1685
New Product
Leverandør:   VWR Collection
VWR® Socket Caps and Colour Inserts for Screw Microtubes
Katalog # 525-1685
Leverandør:  VWR Collection
New Product
VWR® Socket Caps and Colour Inserts for Screw Microtubes
VWR® Socket Caps and Colour Inserts for Screw Microtubes
Customisable screw caps, non sterile
Price
430,00 kr
Log ind for tilgængelighed
1 * 500 Items
Product Details & Documents
Customisable socket caps with O-ring, accept colourful cap inserts to organise samples or match kit branding. For use with VWR® screw cap microtubes only.
Feature ¾ turn to easily open and close tubes requiring less revolutions than competitor tubes.
Specifikationer
  • Beskrivelse:
    Customisable screw caps, non sterile
  • Seq:
    1