Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
VWR® Screw Cap Microtubes with Socket Caps

VWR® Screw Cap Microtubes with Socket Caps

Freestanding microtubes with clear tethered, socket cap, no graduations, combo pack, 0,5 ml, klar, -
Katalog # 525-1653
New Product
Leverandør:   VWR Collection
VWR® Screw Cap Microtubes with Socket Caps
Katalog # 525-1653
Leverandør:  VWR Collection
New Product
VWR® Screw Cap Microtubes with Socket Caps
VWR® Screw Cap Microtubes with Socket Caps

Freestanding microtubes with clear tethered, socket cap, no graduations, combo pack, 0,5 ml, klar, -

Price
674,00 kr
Log ind for tilgængelighed
1 * 500 Items
Product Details & Documents

VWR® screw cap microtubes - Secure storage for valuable samples. VWR® screw cap microtubes provide ultra-pure polypropylene construction, leak-proof caps, and sterile/non sterile options for reliable lab sample storage.

  • Ultra-pure polypropylene (PP): Ensures chemical resistance and compliance with FDA 21 CFR 177.1520 and USP Class VI standards

  • High-clarity design: Crystal-clear walls for easy sample visibility
  • Secure O-ringed flat caps: ³/₄ turn design for leak-proof sealing and convenient opening/closing
  • Designed with an insertable ridge for convenient sample coding using colourful insert chips
  • Versatile temperature performance: Stable from −196 to +121 °C (not recommended for liquid nitrogen immersion)
  • Sterile and non sterile options: Choose pre-sterilised assemblies or convenient non sterile combo packs
  • Multiple formats: Available in the three most commonly used volumes, with conical or skirted/free-standing bottoms for different applications

    Maximum RCF: 20000×g

VWR® screw cap microtubes provide reliable, contamination-free storage for your critical samples, reagents, and biological materials. Designed for durability and clarity, these high-quality tubes are trusted in life science, clinical, and research laboratories worldwide. Ideal for sample storage, molecular biology, PCR, centrifugation, and general lab use.

Certifications: Certified free of detectable DNase, RNase, DNA, and PCR inhibitors. Sterile items are also certified free of detectable endotoxins/pyrogens. Manufactured in compliance with stringent laboratory quality standards.

Bestillingsoplysninger: Spare caps and colour insert chips also available.

Documents

Specifikationer
  • Steril:
    -
  • Kapacitet:
    0,5 ml
  • Farve:
    klar
  • Beskrivelse:
    Freestanding microtubes with clear tethered, socket cap, no graduations, combo pack
  • Seq:
    15