MPC Coupling Caps
SaniQuik USP class VI, ADCF silicone seal, ¾", 100 pk, silicone, Silicone
Katalog # MFLX30003-00
New Product
Leverandør: Avantor Fluid Handling
Gå til:
Product Details & Documents
MPC connectors are multi-purpose, open-format connectors for use in a wide variety of critical fluid handling applications. This particular type of connector is perfect for smaller bag systems for aliquoted media or other products stored in bags.
Ergonomic thumb latch: Easy to operate - even with gloved hands
- USP Class VI materials: Meet biocompatibility
- Sterilisable by autoclave, EtO, e-beam, or gamma: Robust and economical for single-use applications
- Parting line-free hose barb: Eliminates potential leak path
- BSE/TSE free materials: Meet ADCF requirements
MPC connectors add ease of use and security to the most critical fluid handling applications. Choose from a full line of male and female MPC connectors and configurations, including pressure sealing caps and plugs, in sizes to fit ¹/₈", ¹/₄", and ³/₈" tubing. MPC couplings offer optional locking sleeves to further guard against accidental disconnects. In addition, coupling halves can be rotated when connected to reduce tube kinks.
Specifikationer
- Beskrivelse:SaniQuik USP class VI, ADCF silicone seal, ¾", 100 pk, silicone
- Seq:9
- Fitting material:Silicone