Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Immunotag™ Human Complement factor B ELISA Kit

Immunotag™ Human Complement factor B ELISA Kit

Complement factor B ELISA kit
Katalog # GENOIT16410
Leverandør:   G-Biosciences
Immunotag™ Human Complement factor B ELISA Kit
Katalog # GENOIT16410
Leverandør:  G-Biosciences
CAS nummer:  
Immunotag™ Human Complement factor B ELISA Kit

Complement factor B ELISA kit

CAS nummer:  
Price
8.950,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Ready-to-use enzyme-linked immunosorbent assay kits (ELISA Kits).

  • Size: 96 well plate

Complement factor B ELISA kit is based on a sandwich enzyme-linked immunosorbent assay (ELISA) technology. A 96-well plate is coated with a specific antibody against the protein of interest, which will bind the protein and this is detected by a biotin conjugated antibody against the protein of interest. The biotin moiety is subsequently detected, following washing, by the addition of streptavidin coupled horseradish peroxidase (HRP). The supplied TMB substrates were used to visualise HRP enzymatic reaction. TMB was catalysed by HRP to produce a blue color product that changed into yellow after adding acidic stop solution. The density of yellow is directly proportional to the concentration of protein of interest captured on the plate.

Specifikationer

Specifikationer

  • Beskrivelse:
    Complement factor B ELISA kit
  • Cat. No.:
    GENOIT16410
  • Sensitivitet:
    0,297 ng/Ml
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Detektionsområde:
    0,625 - 40 ng/ml
  • Target protein:
    CFB
  • Opbevaringstemperatur:
    Refrigerate (2…8 °C)
  • Regulatory status:
    RUO
  • Analysetype:
    Sandwich

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.