Kinetex Bulk Packing
New Product
Leverandør: PHENOMENEX
Gå til:
Om denne vare
14 tilgængelige muligheder nedenfor
Product Details & Documents
Product Family Options
Produktinformation
- PartikelstørrelsePorestørrelseTilgængelighedPrice
- PHEC04A-4461HILICSpecifikationer:New ProductNew Product
- PHEC04A-4462C18Specifikationer:New ProductNew Product
- PHEC04A-4474HILICSpecifikationer:New ProductNew Product
- PHEC04A-4475C18Specifikationer:New ProductNew Product
- PHEC04A-4476PFPSpecifikationer:New ProductNew Product
- PHEC04A-4477PFPSpecifikationer:New ProductNew Product
- PHEC04A-4497C8Specifikationer:New ProductNew Product
- PHEC04A-4498XB-C18Specifikationer:New ProductNew Product
- PHEC04A-4499C8Specifikationer:New ProductNew Product
- PHEC04A-4601C18Specifikationer:New ProductNew Product
- PHEC04A-4602PFPSpecifikationer:New ProductNew Product
- PHEC04A-4603Phenyl-HexylSpecifikationer:New ProductNew Product
- PHEC04A-4605XB-C18Specifikationer:New ProductNew Product
- PHEC04A-4726EVO C18Specifikationer:New ProductNew Product