trans-Anethole
trans-Anethole
EHERC10256000
- SMILES: COc1ccc(\C=C\C)cc1
- Inchi: InChI=1S/C10H12O/c1-3-4-9-5-7-10(11-2)8-6-9/h3-8H,1-2H3/b4-3+
- IUPAC: 1-methoxy-4-[(E)-prop-1-enyl]benzene
- Packing group: III
- Beskrivelse:trans-Anethole
- Cat. No.:EHERC10256000
- UN No.:3082
- CAS-nr:4180-23-8
- Molekylvægt:148,2
- Fareklassebeskrivelse:GHS07, GHS09
- Opbevaring betingelser:20 °C
- GHS signal:Advarsel
- Synonym(s):trans-Anethole;Benzene, 1-methoxy-4-(1E)-1-propen-1-yl-;Anisole, p-propenyl-, (E)- (8CI);Benzene, 1-methoxy-4-(1-propenyl)-, (E)-;Benzene, 1-methoxy-4-(1E)-1-propenyl- (9CI);1-Methoxy-4-(1E)-1-propen-1-ylbenzene;(E)-1-(4-Methoxyphenyl)propene;(E)-1-p-Methoxyphenylpropene;(E)-Anethol;(E)-Anethole;1-Methoxy-4-[(1E)-1-propenyl]benzene;NSC 209529;trans-1-(4-Methoxyphenyl)-1-propene;trans-1-(p-Methoxyphenyl)-1-propene;trans-1-(p-Methoxyphenyl)propene;trans-1-p-Anisylpropene;trans-4-(1-Propenyl)anisole;trans-Anethol;trans-p-Anethole;trans-p-Methoxy-beta-methylstyrene
- Shipping conditions:RT
- Formel:C₁₀H₁₂O
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



