Saccharin
Saccharin
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: O=C1NS(=O)(=O)c2ccccc12
- Inchi: InChI=1S/C7H5NO3S/c9-7-5-3-1-2-4-6(5)12(10,11)8-7/h1-4H,(H,8,9)
- IUPAC: 1,1-dioxo-1,2-benzothiazol-3-one
Documents
Certifikater
Søg efter partinummerSpecifikationer
Specifikationer
- Beskrivelse:Saccharin
- Cat. No.:EHERC16901000
- CAS-nr:81-07-2
- Molekylvægt:183,18
- Opbevaring betingelser:20 °C
- Synonym(s):Saccharin;1,2-Benzisothiazol-3(2H)-one 1,1-dioxide;1,2-Benzisothiazolin-3-one, 1,1-dioxide (8CI);1,1-Dioxido-3-oxo-2,3-dihydrobenzo[d]isothiazole;1,1-Dioxo-1,2-benzisothiazol-3(2H)-one;1,1-Dioxo-1,2-benzothiazol-3-one;1,2-Dihydro-2-ketobenzisosulfonazole;2,3-Dihydro-1,2-benzisothiazol-3-one-1,1-dioxide;2,3-Dihydro-3-oxobenzisosulfonazole;3-Benzisothiazolinone 1,1-dioxide;3-Hydroxybenzisothiazole-S,S-dioxide;550 Saccharine;Anhydro-o-sulfaminebenzoic acid;Benzoic sulfimide;Benzoic sulphinide;Benzosulfimide;Benzosulfinide;Garantose;Glucid;Gluside;NSC 5349;Necta Sweet;Saccharimide;Saccharin;Saccharin acid;Saccharin insoluble;Saccharine;Saccharinol;Saccharinose;Saccharol;o-Benzoic acid sulfimide;o-Benzoic sulfimide;o-Benzosulfimide;o-Benzoyl sulfimide;o-Sulfobenzimide;o-Sulfobenzoic acid imide
- Shipping conditions:RT
- Formel:C₇H₅NO₃S
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



