Flugestone acetate
Flugestone acetate
Katalog # EHERC13717200
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: CC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)C
- Inchi: InChI=1S/C23H31FO5/c1-13(25)22(29-14(2)26)10-8-17-18-6-5-15-11-16(27)7-9-20(15,3)23(18,24)19(28)12-21(17,22)4/h11,17-19,28H,5-10,12H2,1-4H3/t17-,18-,19-,20-,21-,22-,23-/m0/s1
- IUPAC: [(8S,9R,10S,11S,13S,14S,17R)-17-acetyl-9-fluoro-11-hydroxy-10,13-dimethyl-3-oxo-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl] acetate
- Packing group: III
Specifikationer
Specifikationer
- Beskrivelse:Flugestone acetate
- UN No.:3077
- Cat. No.:EHERC13717200
- CAS-nr:2529-45-5
- Molekylvægt:406,49
- Fareklassebeskrivelse:GHS08, GHS09
- Opbevaring betingelser:4 °C
- GHS signal:Farlig
- Synonym(s):Pregn-4-ene-3,20-dione, 9-fluoro-11β,17-dihydroxy-, 17-acetate (7CI,8CI);Progesterone, 9-fluoro-11β,17-dihydroxy-, 17-acetate (6CI);(11β)-17-(Acetyloxy)-9-fluoro-11-hydroxypregn-4-ene-3,20-dione;17α-Acetoxy-9α-fluoro-11β-hydroxy-pregn-4-ene-3,20-dione;17α-Acetoxy-9α-fluoro-11β-hydroxyprogesterone;9α-Fluoro-11β-hydroxy-17α-acetoxypregn-4-ene-3,20-dione;Chronogest;Cronolone;FGA;Flugestone acetate;Fluorogestone acetate;Flurogestone acetate;Gyncro-Mate;NSC 65411;SC 9880;Synchronate;Syncro-Mate
- Shipping conditions:RT
- Formel:C₂₃H₃₁FO₅



