Dichlorprop-P
Dichlorprop-P
Katalog # EHERC12510500
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: C[C@@H](Oc1ccc(Cl)cc1Cl)C(=O)O
- Inchi: InChI=1S/C9H8Cl2O3/c1-5(9(12)13)14-8-3-2-6(10)4-7(8)11/h2-5H,1H3,(H,12,13)/t5-/m1/s1
- IUPAC: (2R)-2-(2,4-dichlorophenoxy)propanoic acid
Specifikationer
Specifikationer
- Beskrivelse:Dichlorprop-P
- Cat. No.:EHERC12510500
- CAS-nr:15165-67-0
- Molekylvægt:235,06
- Fareklassebeskrivelse:GHS05, GHS07
- Opbevaring betingelser:20 °C
- GHS signal:Advarsel
- Synonym(s):Propanoic acid, 2-(2,4-dichlorophenoxy)-, (2R)-;Propanoic acid, 2-(2,4-dichlorophenoxy)-, (R)-;Propionic acid, 2-(2,4-dichlorophenoxy)-, (+)- (8CI);(2R)-2-(2,4-Dichlorophenoxy)propanoic acid;(+)-2,4-DP;(+)-2-(2,4-Dichlorophenoxy)propanoic acid;(+)-2-(2,4-Dichlorophenoxy)propionic acid;(+)-Dichlorprop;(R)-(+)-2-(2,4-Dichlorophenoxy)propionic acid;(R)-2-(2,4-Dichlorophenoxy)propanoic acid;(R)-2-(2,4-Dichlorophenoxy)propionic acid;(R)-Dichloroprop;(R)-Dichlorprop;Dichlorprop-P;Duplosan DP;R-(+)-Dichlorprop
- Shipping conditions:Room temperature
- Formel:C₉H₈Cl₂O₃
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



