Coumaphos
Coumaphos
Katalog # EHERC11730000
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- SMILES: CCOP(=S)(OCC)Oc1ccc2C(=C(Cl)C(=O)Oc2c1)C
- Inchi: InChI=1S/C14H16ClO5PS/c1-4-17-21(22,18-5-2)20-10-6-7-11-9(3)13(15)14(16)19-12(11)8-10/h6-8H,4-5H2,1-3H3
- IUPAC: 3-chloro-7-diethoxyphosphinothioyloxy-4-methylchromen-2-one
- Packing group: II
Specifikationer
Specifikationer
- Beskrivelse:Coumaphos
- UN No.:2783
- Cat. No.:EHERC11730000
- CAS-nr:56-72-4
- Molekylvægt:362,77
- Fareklassebeskrivelse:GHS06, GHS09
- Opbevaring betingelser:4 °C
- GHS signal:Farlig
- Synonym(s):Coumaphos;Coumaphos
- Shipping conditions:RT
- Formel:C₁₄H₁₆ClO₅PS
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



